Cleanroom shoes, clogs, Hygiene
Supplier: RODO GROUP
Certificates
About this item
Safety clogs made from water resistant microfibre upper. Self cleaning, lightweight PU sole (resistant to oils, fats, acids and alkalis, food and animal waste).
- Anti-static, flexible sole with high slip resistance
- Breathable and fast drying mesh lining
- Foldable, elasticated heel strap
- Machine washable at 40 °C
- Protective steel toecap (200 J)
Specially designed for cleanrooms, medical and food industries.
Colour: White
Certifications: EN 20345; S2, SRC
Frequently Bought Together