Order Entry
Ireland
ContactUsLinkComponent
Autoclave indicator tape
Autoclave indicator tape
Catalog # 116-0426
Supplier:  Synergy Health
undefined
Autoclave indicator tape
Catalog # 116-0426
Supplier:  Synergy Health

Specifications

  • Width:
    24 mm
  • Description:
    Autoclave indicator tape
  • Length:
    50 mm
  • Seq:
    02
  • Pk:
    36

Specifications

About this item

Crepe paper coated with a dry natural rubber based adhesive for steam sterilisation.

  • Good adhesion on wrap made with cellulose fibres
  • Good resistance to sterilisation process
  • Indicator colour change: from green to brown/black

Products must be stored in their original packaging, in dry and cool areas away from humidity.
It’s recommended to use the tape within two years after manufacturing.

Certifications: Conformity ISO 11140-1, CE Mark.