Order Entry
Ireland
ContactUsLinkComponent
Surface cleaner for stainless steel
Surface cleaner for stainless steel
Catalog # MMMAEDEL600
Supplier:  3M
undefined
Surface cleaner for stainless steel
Catalog # MMMAEDEL600
Supplier:  3M
Specifications
  • Description:
    Stainless steel cleaner
  • Environmentally Preferable:
  • Packed:
    Spray can
  • Pk:
    600 mL
Specifications
About this item
Spray cleaner for a stainless steel but can be also used on a range of surfaces including chrome, brass, aluminium, anodised metal surfaces, plastic and enamel. Cleans and protects stainless steel surfaces in one operation, prevents oxidation and fingerprints.