Microcentrifuges, ventilated/refrigerated, 1-16 / 1-16K
: SIGMA
This laboratory microfuge is the replacement for the popular Sigma 1-15 microcentrifuge. The 1-16 centrifuge has a higher max RCF of 20627 ×g.
- Bright display with big push buttons
- Minimal temperature increase during run
- Maintenance-free brushless drive motor, motorised lid lock, auto lid open, emergency lid lock release
- Two acceleration- and deceleration curves, soft or fast
- Window in the lid for external speed control
- 1-16K only: Rotor precooling during standstill, refrigerant CFC-free (R 134 a)
- 1-16K only: Guaranteed temperature of 4 °C at max speed for all rotors (1-16K)
Greater programmability allows the user to specify start and stop accelerations to best serve material being separated. Ideal for sedimentation in microtubes.
: Centrifuges are supplied without rotor which needs to be ordered separately.
Frequently Bought Together