Order Entry
Ireland
ContactUsLinkComponent
Tubing connectors, stepped, SP Bel-Art
Tubing connectors, stepped, SP Bel-Art
  BELAH195630000
undefined
Tubing connectors, stepped, SP Bel-Art
  BELAH195630000
  • Tubing int.Ø:
    9,5 - 12,7 mm
  • Length:
    44,5/14,3 mm
  • Pk:
    12
PP, translucent.
  • Excellent chemical resistance for use with a wide range of liquids or gases
  • Excellent temperature resistance, autoclavable
  • FDA-grade for direct contact with food, drugs, and cosmetics
These fittings ensure a tight, secure connection between tubing having different inner diameters (Int. Ø.).