Centrifuges, benchtop, vent./heat./refrig., 6-16S / 6-16HS / 6-16KS / 6 - 16KHS
: SIGMA
This universal centrifuge can be used with various fixed-angle or swing-bucket rotors. It is ideal for for low speeds e.g. for cell sedimentation.
- Maintenance-free induction drive motor; microcontroller controls speed, time, temperature and gravitational field
- Two motorised lid locks
- Control unit: Spincontrol S
- Magnetic rotor identification prevents the rotor from overspeeding
- No need to open the casing for emergency lid lock release
- Window in the centrifuge lid for external speed control
- 6-16KHS: Max. rotor temperature 40 - 60 °C depending on rotor and speed
Available as ventilated, refrigerated, heated and refrigerated/heated version.
: Supplied without rotor. Please contact your local Avantor office for further information.
Frequently Bought Together