Order Entry
Ireland
ContactUsLinkComponent
Air sampler, Spin Air
undefined
Air sampler, Spin Air
  • Catalog No:
  • 710-0357
  • 710-0356
  • Model:
  • Spin Air Slave
  • Spin Air
  • Flow rate:
  • 100 L/min
  • 100 L/min
  • Sampling head:
  • Aluminium
  • Aluminium
  • Battery type:
  • Ni Metal Hydride, 7,2 V
  • Ni Metal Hydride, 7,2 V
  • Weight:
  • 1,3 kg
  • 1,7 kg
Frequently Bought Together