Order Entry
Ireland
ContactUsLinkComponent
 
Flasks, flat bottom, narrow neck, with beaded rim
  10/10000
Flasks, flat bottom, narrow neck, with beaded rim
  10/10000

 

  • Ext.Ø:
    279 mm
  • Capacity:
    10000 ml
  • Height:
    400 mm
  • Seq:
    01
  • Neck Ø ext.:
    65 mm
  • Pk:
    1

 

 

SIMAX® borosilicate glass 3,3.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

Marked with capacity and a white labelling area.