Glove liners, BO500W
Supplier: Showa
Certificates
About this item
These seamless knitted nylon gloves with elasticated cuffs feature a PU palm coating. The coating protects the hand from oils and abrasion. Ideal for light assembly tasks where a good grip is needed.
- Lightweight, elastic, low-linting
- Anatomical shape, resistant to deformation
- Breathable back of the hand reduces perspiration
- Minimal allergy risk, no irritation by seams
Suitable industries include agriculture, logistics and construction.
Colour: Black
EN 388 (3131); Cat. II
Certifications: EN 388:2016 (3131X); Cat. II
Specifications
- Catalog No:
- 112-5983
- 112-5982
- 112-5985
- 112-5984
- 112-5986
- Model:
- BO500W
- BO500W
- BO500W
- BO500W
- BO500W
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Nylon
- Nylon
- Nylon
- Nylon
- Nylon
- Finish:
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Palm and fingertips coated
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)
- EN 388:2016 (3131X)