Flasks acc. to Kjeldahl with standard ground joint
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass 3,3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With labelling area.
Frequently Bought Together