Adapters, socket/cone/tubing connections
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Adapters, socket to cone, T-connection with plastic screw thread tubing connector.
Frequently Bought Together