Erlenmeyer flasks with screw cap
Supplier: SMITH SCIENTIFIC
Certificates
About this item
Borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and a white labelling area.
Frequently Bought Together