About this item
The HTS Transwell-96 permeable support has an array of 96-wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
- Choice of either polyester (PET) membrane (0,4 µm, 1,0 µm, and 8,0 µm pore sizes) or polycarbonate (PC) membrane (0,4 µm, 3,0 µm, and 5,0 µm pore sizes)
- 0,143 cm² membrane area per well, providing 20% to 50% more surface area for cell growth than other commercially available systems
- Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access
- Optimised for automation, with multichannel feeder ports, improved gripping surface, and standard bar codes
- The reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabiliser to reduce the risk of spills during handling
- The receiver plate isolates each well to enable 96 individual assays
- Sterile
The HTS Transwell® 24-well permeable support has an array of 24 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 24 Transwell® supports to be handed in a single unit.
HTS Transwell® 96-well permeable support systems and plates The HTS Transwell® 96-well permeable support has an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit. - Choice of either polyester (PET) membrane (1,0 µm or 8,0 µm pore sizes) or PC membrane (0,4 µm, 3,0 µm or 5,0 µm pore sizes) - Large apical and basolateral access ports allow efficient media sampling and facilitate automated or manual access - Optimised for automation, with multichannel feeder ports, improved gripping surface, and standard barcodes - Reservoir plate allows for simultaneous feeding of 96 wells and comes with a removable media stabiliser to reduce the risk of spills during handling - Receiver plate isolates each well to enable 96 individual assays
The HTS Transwell® 96-well permeable support has an array of 96 wells with membrane inserts connected by a rigid, robotics-friendly tray that enables all 96 inserts to be handled as a single unit.
Specifications
- Catalog No:
- 734-2689
- 734-4076
- 734-4114
- 734-4077
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk
- RECs, ISO 14001 - Kennebunk