Tubing, silicone, platinum cured
Supplier: SF MEDICAL
Certificates
About this item
Silicone, platinum cured.
- Extended post cure for lowest level extractables
- Biocompatible, inert, non-reactive with other elements per systemic testing
- Odourless, tasteless, non-toxic for use in medical, food, drug, personal care, and deionised water applications
- Non-leaching plasticisers
- Sterilizable by radiation, ETO, steam –30 bar or mbar at 253 °F (123 °C) for use in sterile applications, reusability
- Colour stripe, tint for custom process control identification
Temperature resistance:—54 to +204 °C
Hardness (Shore durometer A): 60
Density: 1,15
Frequently Bought Together