Accessories for the Dilumat™ START and Dilumat™ EXPERT
Supplier: bioMérieux
Certificates
About this item
Frequently Bought Together
Assistent Brushes with Cotton Tip Bristles
Supplier: GLASWARENFABRIK KARL HECHT
DURAN® protect+ Laboratory Bottles, Plastic Safety Coated, DIN 168-1 Thread, Graduated, Clear
Supplier: DWK Life Sciences
Pipette tips, Finntip™
Supplier: Thermo Fisher Scientific
VWR®, Centrifuge Tubes, Conical Bottom
Supplier: VWR Collection
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Ethanol absolute ≥99.5% USP, Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol 96% (v/v), AnalaR NORMAPUR® analytical reagent
Supplier: VWR Chemicals