Order Entry
Ireland
ContactUsLinkComponent
Vibration spatula
Vibration spatula
Catalog # HAMMHSN462-22
Supplier:  HAMMACHER KARL
undefined
Vibration spatula
Catalog # HAMMHSN462-22
Supplier:  HAMMACHER KARL
Specifications
  • Length:
    220 mm
  • Pk:
    1
Specifications
About this item
Vibration spatula with acid-proof plastic handle, PTFE-coated. For fine dosing.