Fmoc-Osu (N-(9-fluorenylmethoxycarbonyloxy)succinimide)
Supplier: Cayman Chemical
Certificates
About this item
Synonyms: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
CAS Number:82911-69-1
:
Formula:C₁₉H₁₅NO₅
MDL Number:MFCD00010733
Molecular Weight:337.3 g/mol
EINECS:433-520-5
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together