Order Entry
Ireland
ContactUsLinkComponent
 
n-Hexadecyl-β-D-maltoside ≥98%
Catalog # J66531.ME
CAS Number:  
n-Hexadecyl-β-D-maltoside ≥98%
Catalog # J66531.ME
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :566.72 g/mol
Storage Temperature:Freezer
CAS Number:98064-96-1
Formula:C₂₈H₅₄O₁₁
MDL Number:MFCD01310880