
Terpineol, mixture of isomers ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:8000-41-7
:
Formula:C₁₀H₁₈O
MDL Number:MFCD00166983
:154.25 g/mol
Boiling Pt:213…218 °C
Melting Pt:31…36 °C
Density:0,934
Flash Pt:91 °C (196 °F)
EINECS:232-268-1
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together