Toluene, extra dry 99.85%, AcroSeal®
Catalog # 448401000.
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
- Seq:2320
- Pk Info:4× 25 ml
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Water:≤0.003 % (Coulometric)
- GC:≥99.8 %
- Residue after evaporation:≤0.0005 %
- Filt.over 0.2 micron PFTE:Filtered
Specification Test Results
About this item
:
:92.14 g/mol
Merck Index:13,09607
Boiling Pt:111 °C (760 mmHg)
Melting Pt:–95 °C
Density:0,866 g/cm³
Flash Pt:4 °C
Storage Temperature:Ambient
EINECS:203-625-9
UN:1294
ADR:3,II
REACH:05-2114615130-69
:GHS07
:GHS08
:GHS02
CAS Number:108-88-3
Formula:C₆H₅CH₃
MDL Number:MFCD00008512