0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
Sodium chloride
Sodium chloride 0,85 - 0,9% in aqueous solution
Pk:
4 L
Seq:
5162
Environmentally Preferable:
:
Saline, NaCl, Sea salt, Halite
:
:
58.44 g/mol
:
Ambient
:
231-598-3
:
7647-14-5
:
NaCl
:
MFCD00003477