Ethyl-3-piperidinecarboxylate ≥97%
Catalog # B24155.14
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
CAS Number:5006-62-2
:
Formula:C₈H₁₅NO₂
MDL Number:MFCD00005991
:157.21 g/mol
Boiling Pt:102…104 °C (7 mmHg)
Density:1,012
Flash Pt:90 °C (195 °F)
EINECS:225-681-3
:GHS07