Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Cyano-2-methylpyridine
5-Cyano-2-methylpyridine
5-Cyano-2-methylpyridine
MAYBSPB05015.1
:
Thermo Scientific
:
5-Cyano-2-methylpyridine
MAYBSPB05015.1
:
Thermo Scientific
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
118.14 g/mol
:
3222-48-8
:
C₇H₆N₂
:
MFCD00038042