
Calmagite, pure indicator grade
:3147-14-6
:
:C₁₇H₁₄N₂O₅S
:MFCD00011656
:358.37 g/mol
:300 °C
:221-563-0
:GHS07
:Ambient
- Appearance:Black Crystalline powder
- Infrared spectrum:Conforms
- Specific extinction E (1%/1cm):≥300 (610 nm, in buffer)
- Suitability:(for Ca + Mg determination) Passes test
- UV:(2.7 g in 100 ml water) pH 3
- Titration after Ion exchange:≥99.0 %
Frequently Bought Together