Order Entry
Ireland
Orders LinkContactUsLinkComponent
PTFE/rubber laminated discs
PTFE/rubber laminated discs
Certificates
PTFE/rubber laminated discs
PTFE/rubber laminated discs
3 Options Available Below
Product Details & Documents
These discs are designed to provide a highly inert and unreactive seal. Constructed of white pharmaceutical rubber with 0,1 mm of PTFE coating bonded to one side, they are excellent for use as cap liners when a highly inert non reactive seal is desired.
  • Autoclavable with no loss of integrity after heating above 100 °C for five hours
Discs are rigid and more difficult to puncture than PTFE/silicone discs.
Product Family Options
Product Information
  • For
    Ø
    Thickness
    Availability
    Price
  • HYPETS-12412
    PTFE/rubber laminated disc
    PTFE/rubber laminated disc
    0,3 and 1,0 ml Reacti-Vial™ small reaction vials, 2 and 3,5 ml screw cap septum vials
    12 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items - 101,00 €
    101,00 €
    1 * 72 items
       101,00 €
  • HYPETS-12418
    PTFE/rubber laminated disc
    PTFE/rubber laminated disc
    3 and 5 ml Reacti-Vial™ small reaction vials; 10 and 25 ml Reacti-Flask
    18 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items - 113,00 €
    113,00 €
    1 * 72 items
       113,00 €
  • HYPETS-12422
    PTFE/rubber laminated disc
    PTFE/rubber laminated disc
    25 and 40 ml screw cap septum vials
    22 mm
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 72 items
    1 * 72 items - 128,00 €
    128,00 €
    1 * 72 items
       128,00 €