HPLC columns, HyPURITY™ Aquastar
Supplier: Thermo Fisher Scientific
Certificates
About this item
These columns are a silica-based polar end-capped and it is designed to provide superior retention of polar compounds and increased sensitivity in 100% aqueous conditions.
- Ideal for structurally similiar analytes
- Optimised for LC/MS applications
Frequently Bought Together