Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Safety Glasses
Pouch for Raptor safety spectacles
Pouch for Raptor safety spectacles
Pouch for Raptor safety spectacles
Catalog # CENUS70POUCH
Supplier:
CENTURION SAFETY PRODUCTS
Expand image
Pouch for Raptor safety spectacles
Catalog # CENUS70POUCH
Supplier:
CENTURION SAFETY PRODUCTS
Specifications
Description:
Storage pouch for safety spectacles Raptor, Centurion
Pk:
1
Specifications
About this item
Product Details
Product Details