
4-[4-(Trifluoromethoxy)phenoxy]piperidine ≥97%
Catalog # H50812.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:287952-67-4
:
Formula:C₁₂H₁₄F₃NO₂
MDL Number:MFCD06656166
:261.25 g/mol
Melting Pt:71…73 °C
:GHS07
Storage Temperature:Ambient