Order Entry
Ireland
ContactUsLinkComponent
Nitric acid 0.1 mol/l (0.1 N), Supelco®
Corrosive
Nitric acid 0.1 mol/l (0.1 N), Supelco®
Catalog # 16355-1L
Supplier:  Merck
CAS Number:  
undefined
Corrosive
Nitric acid 0.1 mol/l (0.1 N), Supelco®
Catalog # 16355-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :63.01 g/mol
ADR:8,III
 :M-Clarity™ quality level = MQ100
EINECS:231-714-2
UN:3264
 :GHS05
Storage Temperature:Ambient
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349