Order Entry
Ireland
ContactUsLinkComponent
2-Chloro-7-methoxy-4-methylquinoline ≥97%
Irritant
2-Chloro-7-methoxy-4-methylquinoline ≥97%
Catalog # H32685.03
CAS Number:  
Irritant
2-Chloro-7-methoxy-4-methylquinoline ≥97%
Catalog # H32685.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:97892-67-6
 :
Formula:C₁₁H₁₀ClNO
MDL Number:MFCD00711757
 :207.66 g/mol
Melting Pt:111…113 °C
 :GHS07
Storage Temperature:Ambient