Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3,4-Trichloroquinoline
2,3,4-Trichloroquinoline
2,3,4-Trichloroquinoline
2,3,4-Trichloroquinoline
Catalog # MAYBBTB10984.1
Supplier:
Thermo Scientific
CAS Number:
Expand image
2,3,4-Trichloroquinoline
Catalog # MAYBBTB10984.1
Supplier:
Thermo Scientific
CAS Number:
Specifications
Pk:
1 g
Seq:
3047
Environmentally Preferable:
Specifications
About this item
Product Details
:
CAS Number:
40335-02-2
Formula:
C₉H₄Cl₃N
Product Details