Order Entry
Ireland
ContactUsLinkComponent
Crystallisation plates, CrystalStar™, 1536-well IMP@CT™
Crystallisation plates, CrystalStar™, 1536-well IMP@CT™
Catalog # 737-3507
Supplier:  Greiner Bio-One
undefined
Crystallisation plates, CrystalStar™, 1536-well IMP@CT™
Catalog # 737-3507
Supplier:  Greiner Bio-One
Specifications
  • Description:
    1536-well microplate lid, low profile (6 mm), non sterile
  • Seq:
    04
  • Pk:
    200
Specifications
About this item
Designed for microbatch applications under oil, the IMP@CT™ crystallisation plates are characterised by conical wells with a flat bottom.