Connectors, tubing, KECK™
Supplier: Roth Carl
Certificates
About this item
Tubing connectors made of PBT with EPDM flat seal. Ideal for fast, safe and leakproof connections of tubing to glass hose connectors and glass threads.
- High chemical resistance
- Temperature stable to max. 130 °C
- Suitable for glass olives (clamp, ring, thread cap, tubing sleeve connector, seals)
Frequently Bought Together