Bottles, narrow neck, soda-lime glass, clear or amber, with standard ground glass stopper
Bottles, made of soda-lime glass, with conical ground glass joint and standard ground glass stopper, solid glass (NS 14 to NS 24) or semi-hollow (in NS 29 or larger).
- Good chemical and physical properties
- For use in applications that are not associated with high thermal loads
- Stable and tough
- Amber coloured for light sensitive substances
Frequently Bought Together