Chemical resistant gloves, Alto 415
Supplier: MAPA
Certificates
About this item
These neoprene/natural latex gloves with textured finish and cotton flock lining are suitable for spray-painting, industrial cleaning, maintenance and for work with composite materials in the aeronautics industry.
- Tactile sensitivity, due to the gloves reduced thickness
- Cotton flock-lining esnures comfort and perspiration absorption
- Good grip thanks to the non-slip embossing
Certifications: EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421; Cat. III
Specifications
- Catalog No:
- MAPA34415049
- MAPA34415040
- MAPA34415151
- MAPA34415048
- MAPA34415046
- MAPA34415047
- Model:
- Alto 415
- Alto 415
- Alto 415
- Alto 415
- Alto 415
- Alto 415
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Embossed
- Embossed
- Embossed
- Embossed
- Embossed
- Embossed
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421
- EN 388:2016 (1011X), EN ISO 374-1 Type B (KMT), EN ISO 374-5, EN 421