Industrial gloves, Titan 397
Supplier: MAPA
Certificates
About this item
These nitrile gloves with textile support and smooth finish provide comfort and dexterity for common handling tasks.
- Excellent comfort, due to the thickness of the cotton lining and the ventilated back
- Freedom of movement: dexterity and flexibility, due to the close fit to the hand
- Good abrasion resistance
Ideal for sanding, precision mechanical work, part assembly, handling oily mechanical parts, manufacturing small electrical components and for construction materials handling.
Certifications: EN 388:2016 (4111X); Cat. II
Specifications
- Catalog No:
- 112-5582
- 112-5583
- 112-5579
- 112-5580
- 112-5581
- Model:
- Titan 397
- Titan 397
- Titan 397
- Titan 397
- Titan 397
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- ¾ coating
- ¾ coating
- ¾ coating
- ¾ coating
- ¾ coating
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- EN norm:
- EN 388:2016 (4111X)
- EN 388:2016 (4111X)
- EN 388:2016 (4111X)
- EN 388:2016 (4111X)
- EN 388:2016 (4111X)