Order Entry
Ireland
ContactUsLinkComponent
General purpose gloves, Titan 328
General purpose gloves, Titan 328
Supplier:  MAPA
Certificates
undefined
General purpose gloves, Titan 328
Supplier:  MAPA
Specifications
  • Catalog No:
  • MAPA34328029
  • MAPA34328020
  • MAPA34328028
  • Model:
  • Titan 328
  • Titan 328
  • Titan 328
  • Ambidextrous / hand-specific:
  • Hand-specific
  • Hand-specific
  • Hand-specific
  • Disposable / reusable:
  • Reusable
  • Reusable
  • Reusable
  • Liner material:
  • Cotton
  • Cotton
  • Cotton
  • Finish:
  • Embossed
  • Embossed
  • Embossed
  • Cuff style:
  • Knit wrist
  • Knit wrist
  • Knit wrist
  • EN norm:
  • EN 388:2016 (2142X), EN 407 (X2XXXX)
  • EN 388:2016 (2142X), EN 407 (X2XXXX)
  • EN 388:2016 (2142X), EN 407 (X2XXXX)