Industrial gloves, Ultranitril 472
Supplier: MAPA
Certificates
About this item
These nitrile gloves with chlorinated interior finish and pebbled exterior finish are suitable for use in the food industry, mechanical industry and in the construction industry (carpenters/joiners). They can be worn alone or as an outer-glove.
- Fingertip precision for light chemical protection and food handling
- Freedom of movement and excellent sensitivity, due to the gloves reduced thickness
- Chlorinated finish for better protection of the products being handled
- Suitable for all types of food
Certifications: EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421; Cat. III
Food contact approved
Food contact approved
Specifications
- Catalog No:
- MAPA34472269
- MAPA34472268
- MAPA34472247
- MAPA34472266
- MAPA34472260
- Model:
- Ultranitril 472
- Ultranitril 472
- Ultranitril 472
- Ultranitril 472
- Ultranitril 472
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Finish:
- Pebbled
- Pebbled
- Pebbled
- Pebbled
- Pebbled
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421
- EN 388:2016 (2101X), EN ISO 374-1 Type B (JOT), EN ISO 374-5 VIRUS, EN 421
- Food contact approved:
- +
- +
- +
- +
- +