Chemical resistant gloves, Alto 299
Supplier: MAPA
Certificates
About this item
Natural latex gloves with flocked interior.
- Longer service life and good impact protection, due to the thickness of the material
- Embossed texture for improved grip
- Good mechanical performance for long-lasting chemical protection
Ideal for maintenance tasks in in wet environments (contact with water, oil, greases, hydrocarbons) and for emptying and cleaning septic tanks.
Certifications: EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5; Cat. III
Specifications
- Catalog No:
- MAPA34299180
- MAPA34299187
- MAPA34299189
- MAPA34299188
- Model:
- Alto 299
- Alto 299
- Alto 299
- Alto 299
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Embossed
- Embossed
- Embossed
- Embossed
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5
- EN 388:2016 (3121X), EN ISO 374-1 Type A (AKLMPT), EN ISO 374-5