Chemical resistant gloves, Telsol 351
Supplier: MAPA
Certificates
About this item
PVC gloves with knitted textile support for improved comfort. The gloves provide protection against mechanical and low-risk chemical hazards.
- Exceptional comfort and good resistance to cold, due to the seamless cotton-knit
- Good resistance to oils, greases and hydrocarbons
- Suitable for food contact, except fatty foods
- Pebbled finish
Ideal for use in the pharmaceutical industry and for maintenance and cleaning tasks in wet environments (water, oil, greases, hydrocarbons).
Certifications: EN 388:2016 (4121X), EN ISO 374-1 Type A (KLMNPT), EN ISO 374-5; Cat. III; AQL <0,65.
Specifications
- Catalog No:
- MAPA34351008
- MAPA34351009
- MAPA34351000
- Model:
- Telsol 351
- Telsol 351
- Telsol 351
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Finish:
- Rough
- Rough
- Rough
- Cuff style:
- Gauntlet
- Gauntlet
- Gauntlet
- EN norm:
- EN 388:2016 (4121X), EN ISO 374-1 Type A (KLMNPT), EN ISO 374-5
- EN 388:2016 (4121X), EN ISO 374-1 Type A (KLMNPT), EN ISO 374-5
- EN 388:2016 (4121X), EN ISO 374-1 Type A (KLMNPT), EN ISO 374-5