Industrial gloves, Alto 258
Supplier: MAPA
Certificates
About this item
These liquidproof gloves made of natural latex provide a good resistance to many diluted acids.
- Cotton flock-lining over an internal layer of white natural rubber
- Curved fingers and contoured palm
- Non-slip textured finish in palm and fingers area for improved grip
- External surface lightly coated with silicone
Typical applications include: Work with composite materials (resins), common handling tasks, assembly of small, oil and grease free parts and minor maintenance tasks.
Thickness: 0,60 mm
Colour: Yellow
EN 388 (2120), EN 374; Cat. III; AQL 0,65 Food contact approved
Certifications: EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5; Cat. III
Specifications
- Catalog No:
- 112-2997
- 112-2993
- 112-2998
- 112-2999
- Model:
- Alto 258
- Alto 258
- Alto 258
- Alto 258
- Ambidextrous / hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Cotton
- Cotton
- Finish:
- Textured
- Textured
- Textured
- Textured
- Cuff style:
- Straight
- Straight
- Straight
- Straight
- EN norm:
- EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5
- EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5
- EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5
- EN 388 (1110X), EN ISO 374-1 Type B (KPS), EN ISO 374-5