Order Entry
Ireland
ContactUsLinkComponent
Lead acetate solution Reag. Ph. Eur. 1048103
Lead acetate solution Reag. Ph. Eur. 1048103
Catalog # 87860.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Lead acetate solution Reag. Ph. Eur. 1048103
Catalog # 87860.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
Storage Temperature:Ambient
UN:3082
ADR:9,III
 :GHS08
 :GHS09