Order Entry
Ireland
ContactUsLinkComponent
Iron(III) chloride solution R3 Reag. Ph. Eur. 1037803
Flammable
Irritant
Iron(III) chloride solution R3 Reag. Ph. Eur. 1037803
Catalog # 87835.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Flammable
Irritant
Iron(III) chloride solution R3 Reag. Ph. Eur. 1037803
Catalog # 87835.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :162.21 g/mol
EINECS:231-729-4
UN:1170
ADR:3,II
 :GHS02
 :GHS07
CAS Number:7705-08-0
Formula:FeCl₃
MDL Number:MFCD00011005