Order Entry
Ireland
ContactUsLinkComponent
Antimony(III) chloride 30% (w/v) in chloroform Reag. Ph. Eur. 1007701
Corrosive
Health
Irritant
Environment
Antimony(III) chloride 30% (w/v) in chloroform Reag. Ph. Eur. 1007701
Catalog # 87779.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Health
Irritant
Environment
Antimony(III) chloride 30% (w/v) in chloroform Reag. Ph. Eur. 1007701
Catalog # 87779.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :228.12 g/mol
UN:1760
ADR:8,II
 :GHS05
 :GHS08
 :GHS07
 :GHS09
CAS Number:10025-91-9
Formula:SbCl₃
MDL Number:MFCD00011212