Order Entry
Ireland
ContactUsLinkComponent
Hydrochloric acid brominated Reag. Ph. Eur. 1043507
Corrosive
Irritant
Hydrochloric acid brominated Reag. Ph. Eur. 1043507
Catalog # 87845.290
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Hydrochloric acid brominated Reag. Ph. Eur. 1043507
Catalog # 87845.290
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
ADR:8,I
 :36.46 g/mol
UN:1760
 :GHS05
 :GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324