Order Entry
Ireland
ContactUsLinkComponent
Silicone oil 550 for gas chromatography, Supelco®
Silicone oil 550 for gas chromatography, Supelco®
Catalog # 1.09762.0100
Supplier:  Merck
CAS Number:  
Silicone oil 550 for gas chromatography, Supelco®
Catalog # 1.09762.0100
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • Solubility:
    (20 °C) practically insoluble
  • Flash Point:
    270 °C
  • Density:
    1.07 g/cm3 (20 °C)
  • Ignition Point:
    400 °C DIN 51794
  • Viscosity kinematic:
    208 mm2/s (20 °C)

Specification Test Results

About this item

 :
Melting Pt:< –35 °C
Density:1,07 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Flash Pt:270 °C
Storage Temperature:Ambient
CAS Number:68083-14-7
 :63148-62-9
MDL Number:MFCD00132673