About this item
Designed to fit the Gilson MICROMAN® positive displacement pipette, Capillary Piston Tips are ideal for difficult-to-pipette samples including; viscous, volatile, dense, hot or cold, or hazardous liquids. The tips work like a syringe providing protection against pipette, sample, and user contamination, and produce accurate, reproducible, and precise results. Made of pure polypropylene, their slim fit allows you to use even the narrowest and deepest test tubes.
- Accurate and precise pipetting
- Protection against contamination
- Convenient
Using Capillary Piston Tips with your MICROMAN ensures maximum accuracy and precision and delivers consistent results, even with viscous, volatile, dense, hot or cold, or hazardous liquids such as radioactive samples. No contact between the sample and the pipette protects against contamination and provides protection for the user and the pipette, even with aerosols or corrosive vapors. The tips are made of pure polypropylene and are free of dyes, so there is no risk of chemical contamination from the tips.
Capillary Piston Tips can pipette volumes from 1 µl to 1000 µl. Choose from six different volume ranges. The compact and convenient TIPACK™ packaging contains color-coded racks for easy identification and compliance with Good Laboratory Practice (GLP).