Order Entry
Ireland
ContactUsLinkComponent
Water Guard, arium®
Water Guard, arium®
Catalog # 171-0899
Supplier:  Sartorius
undefined
Water Guard, arium®
Catalog # 171-0899
Supplier:  Sartorius
Specifications
  • Description:
    arium® Water Guard for Ultra pure water systems arium® Basic and arium® Pro
  • Pk:
    1
Specifications
About this item
Arium® Water guard detects water leaks early. That means the lab gets optimal protection against water damage.
  • High-grade material, non-corrosive
  • Optic and acoustic alarm
  • Easy to install
  • Integrated wall holder for solenoid valve
  • Versatile
Leaks are registered by an optical sensor. This automatically closes the inlet line and triggers an acoustic warning signal. In addition, the user can also constantly monitor the system's status on the built-in LED display.