Order Entry
Ireland
ContactUsLinkComponent
Beakers, low form, porcelain
Beakers, low form, porcelain
Catalog # KAVA641323227201
Supplier:  Kavalier
Beakers, low form, porcelain
Catalog # KAVA641323227201
Supplier:  Kavalier

Specifications

  • Ext.Ø:
    80 mm
  • Capacity:
    400 ml
  • Height:
    99 mm
  • Pk:
    1

Specifications

About this item

Porcelain, glazed white.

  • Exceptional chemical resistance
  • Very good thermal resistance
  • Robust, heavy construction

Cylindrical, with spout.