Order Entry
Ireland
ContactUsLinkComponent
Mag-Bind® PX blood RNA 96 kit
Mag-Bind® PX blood RNA 96 kit
Supplier:  OMEGA BIO-TEK
Certificates
Mag-Bind® PX blood RNA 96 kit
Supplier:  OMEGA BIO-TEK

Specifications

  • Application:
  • Method / Format:
  • Refine target molecule:
  • Sample type:
  • Target molecule:
  • Environmentally Preferable:

Specifications

Related Information

Frequently Bought Together